missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methyl-6-nitroaniline, 97%

Catalog No. AAA1431906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1431906 5 g
AAA1431914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1431906 Supplier Thermo Scientific Chemicals Supplier No. A1431906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 77811-44-0 | C7H7BrN2O2 | 231.049 g/mol

It is used in the design and synthesis of CK2 inhibitors and in the docking studies of novel telmisartan-glitazone hybrid analogs for the treatment of metabolic syndrome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the design and synthesis of CK2 inhibitors and in the docking studies of novel telmisartan-glitazone hybrid analogs for the treatment of metabolic syndrome.

Solubility
Slightly soluble in water and soluble in hot methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 77811-44-0
Molecular Formula C7H7BrN2O2
Molecular Weight (g/mol) 231.049
MDL Number MFCD00052919
InChI Key ZXFVKFUXKFPUQJ-UHFFFAOYSA-N
Synonym 4-bromo-2-methyl-6-nitro-phenylamine, 4-bromo-2-methyl-6-nitrobenzenamine, 2-amino-5-bromo-3-nitrotoluene, 4-bromo-6-nitro-o-toluidine, benzenamine, 4-bromo-2-methyl-6-nitro, 4-bromo-2-methyl-6-nitrophenylamine, 4-bromo-2-methyl-6-nitro-aniline, pubchem3813, acmc-1bg8u, ksc496e7t
PubChem CID 522679
IUPAC Name 4-bromo-2-methyl-6-nitroaniline
SMILES CC1=CC(=CC(=C1N)[N+](=O)[O-])Br

Specifications

Melting Point 143°C to 147°C
Quantity 5 g
Beilstein 3530524
Solubility Information Slightly soluble in water and soluble in hot methanol.
Formula Weight 231.06
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methyl-6-nitroaniline
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only