missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxyphenol, 98%

Catalog No. AC441950050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC441950050 5 g Glass bottle
AC441950250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC441950050 Supplier Thermo Scientific Chemicals Supplier No. 441950050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7368-78-7 | C7H7BrO2 | 203.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7368-78-7
Molecular Formula C7H7BrO2
Molecular Weight (g/mol) 203.04
InChI Key WHSIIJQOEGXWSN-UHFFFAOYSA-N
Synonym 4-bromoguaiacol, 5-bromo-2-hydroxyanisole, 4-bromo-2-methoxy-phenol, phenol, 4-bromo-2-methoxy, 2-methoxy-4-bromophenol, 5bromoguaiacol, 5-bromoguaiacol, 5-bromguajacol, pubchem17271, acmc-209orr
PubChem CID 262234
IUPAC Name 4-bromo-2-methoxyphenol
SMILES COC1=C(C=CC(=C1)Br)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 35°C to 39°C
Color Brown to Colorless
Boiling Point 90°C to 92°C (1.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 203.04
Percent Purity 98%
Physical Form Low Melting Solid or Crystalline Solid
Chemical Name or Material 4-Bromo-2-methoxyphenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only