accessibility menu, dialog, popup

UserName

H3COBrHO

CAS RN 7368-78-7

IUPAC Name: 4-bromo-2-methoxyphenol
Synonyms: 4-bromo-2-methoxyphenol
Molecular Weight (g/mol): 203.04
Molecular Formula: C7H7BrO2
InChi Key: WHSIIJQOEGXWSN-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=CC=C1O
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)