missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxyphenol, 98%

Catalog No. AAL0880118
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0880118 50 g
AAL0880104 2 g
AAL0880109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0880118 Supplier Thermo Scientific Chemicals Supplier No. L0880118

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7368-78-7 | C7H7BrO2 | 203.035 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 7368-78-7
Molecular Formula C7H7BrO2
Molecular Weight (g/mol) 203.035
MDL Number MFCD00051937
InChI Key WHSIIJQOEGXWSN-UHFFFAOYSA-N
Synonym 4-bromoguaiacol, 5-bromo-2-hydroxyanisole, 4-bromo-2-methoxy-phenol, phenol, 4-bromo-2-methoxy, 2-methoxy-4-bromophenol, 5bromoguaiacol, 5-bromoguaiacol, 5-bromguajacol, pubchem17271, acmc-209orr
PubChem CID 262234
IUPAC Name 4-bromo-2-methoxyphenol
SMILES COC1=C(C=CC(=C1)Br)O

Specifications

Melting Point 32°C to 37°C
Boiling Point 129°C to 132°C (12 mmHg)
Odor Phenol-like
Quantity 50 g
Beilstein 2045286
Solubility Information Slightly soluble in water.
Formula Weight 203.04
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-methoxyphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only