missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxyaniline 98.0+%, TCI America™

Catalog No. p-7117691
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20271G 1 g
A20275G 5 g
A202725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A20271G Supplier TCI America Supplier No. A20271G
Only null left

Chemical Identifiers

CAS 59557-91-4
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD01204266
InChI Key WRFYIYOXJWKONR-UHFFFAOYSA-N
Synonym 4-bromo-2-methoxy-phenylamine, 2-amino-5-bromoanisole, 4-bromo-o-anisidine, 4-bromo-2-methoxyphenylamine, 4-bromo-2-methoxy-aniline, 2-methoxy-4-bromoaniline, benzenamine, 4-bromo-2-methoxy, pubchem2258, acmc-209meb, 4-bromo-2-methoxy aniline
PubChem CID 459257
IUPAC Name 4-bromo-2-methoxyaniline
SMILES COC1=C(C=CC(=C1)Br)N

Specifications

Melting Point 59°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 202.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-methoxyaniline