accessibility menu, dialog, popup

UserName

CAS RN 59557-91-4

IUPAC Name: 4-bromo-2-methoxyaniline
Synonyms: 4-bromo-2-methoxyaniline
Molecular Weight (g/mol): 202.05
Molecular Formula: C7H8BrNO
InChi Key: WRFYIYOXJWKONR-UHFFFAOYSA-N
SMILES: COC1=C(N)C=CC(Br)=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)