missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-iodophenol 98.0+%, TCI America™

Catalog No. B49571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49571G 1 g
B49575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49571G Supplier TCI America Supplier No. B49571G
Only null left

Chemical Identifiers

CAS 207115-22-8
Molecular Formula C6H4BrIO
Molecular Weight (g/mol) 298.905
MDL Number MFCD09029644
InChI Key UXIULWIJWDJDQD-UHFFFAOYSA-N
PubChem CID 10924532
IUPAC Name 4-bromo-2-iodophenol
SMILES C1=CC(=C(C=C1Br)I)O

Specifications

Melting Point 75°C
Quantity 1 g
Formula Weight 298.91
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-iodophenol