accessibility menu, dialog, popup

UserName

CAS RN 207115-22-8

BrIOH

CAS RN 207115-22-8

IUPAC Name: 4-bromo-2-iodophenol
Synonyms: 4-bromo-2-iodophenol
Molecular Weight (g/mol): 298.91
Molecular Formula: C6H4BrIO
InChi Key: UXIULWIJWDJDQD-UHFFFAOYSA-N
SMILES: OC1=C(I)C=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results