missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-iodoaniline 97.0+%, TCI America™

Catalog No. B31641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31641G 1 g
B31645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31641G Supplier TCI America Supplier No. B31641G

Chemical Identifiers

CAS 66416-72-6
Molecular Formula C6H5BrIN
Molecular Weight (g/mol) 297.921
MDL Number MFCD02093764
InChI Key HHTYEQWCHQEJNV-UHFFFAOYSA-N
Synonym 4-bromo-2-iodo-phenylamine, 4-bromo-2-iodobenzenamine, 2-iodo-4-bromoaniline, benzenamine,4-bromo-2-iodo, benzenamine, 4-bromo-2-iodo, pubchem3784, acmc-1b9hb, 4-bromo-2-iodophenylamine, asischem t54343, 4-bromo-2-iodo-benzenamine
PubChem CID 823934
IUPAC Name 4-bromo-2-iodoaniline
SMILES C1=CC(=C(C=C1Br)I)N

Specifications

Melting Point 70°C
Color Purple-White
Quantity 1 g
UN Number 2811
Formula Weight 297.92
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-iodoaniline