missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorotoluene 97.0+%, TCI America™

Catalog No. p-7119224
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17165G 5 g
B171625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17165G Supplier TCI America Supplier No. B17165G
Only null left

Chemical Identifiers

CAS 51436-99-8
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00013551
InChI Key YZFVUQSAJMLFOZ-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorotoluene, 2-fluoro-4-bromotoluene, benzene, 4-bromo-2-fluoro-1-methyl, toluene,2-fluoro-4-bromo, 4-bromo-2-fluoro-1-methyl-benzene, pubchem1601, acmc-1an2v, 2-fluoro-4-bromo toluene, 4-bromo-2-fluoro-toluene
PubChem CID 171040
IUPAC Name 4-bromo-2-fluoro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)Br)F

Specifications

Color Yellow
Boiling Point 100°C
Quantity 5 g
Formula Weight 189.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-fluorotoluene