missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorophenylacetic acid, 98%

CAS: 114897-92-6 | C8H6BrFO2 | 233.036 g/mol

Supplier:  Thermo Scientific Chemicals H6117606

 View more versions of this product

Catalog No. AAH6117606


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromo-2-fluorophenylacetic acid
C8H6BrFO2
5 g
PNBIYFPZODYMOO-UHFFFAOYSA-N
2-(4-bromo-2-fluorophenyl)acetic acid
14053792
98%
114897-92-6
MFCD09032952
4-bromo-2-fluorophenylacetic acid, 2-4-bromo-2-fluorophenyl acetic acid, 4-bromo-2-fluorophenyl acetic acid, 4-bromo-2-fluoro-phenyl-acetic acid, 2-4-bromo-2-fluoro-phenyl acetic acid, 4-bromo-2-fluorophenylaceticacid, pubchem13742, acmc-1bskq, ksc493s2b, 4-bromo-2-fluoro-phenylacetic acid
C1=CC(=C(C=C1Br)F)CC(=O)O
233.036
233.03
Solid

Chemical Identifiers

114897-92-6
233.036
PNBIYFPZODYMOO-UHFFFAOYSA-N
14053792
C1=CC(=C(C=C1Br)F)CC(=O)O
C8H6BrFO2
MFCD09032952
4-bromo-2-fluorophenylacetic acid, 2-4-bromo-2-fluorophenyl acetic acid, 4-bromo-2-fluorophenyl acetic acid, 4-bromo-2-fluoro-phenyl-acetic acid, 2-4-bromo-2-fluoro-phenyl acetic acid, 4-bromo-2-fluorophenylaceticacid, pubchem13742, acmc-1bskq, ksc493s2b, 4-bromo-2-fluoro-phenylacetic acid
2-(4-bromo-2-fluorophenyl)acetic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only