missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorophenylacetic acid, 98%

Catalog No. p-7053613
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6117606 5 g
AAH6117614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6117606 Supplier Thermo Scientific Chemicals Supplier No. H6117606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 114897-92-6 | C8H6BrFO2 | 233.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114897-92-6
Molecular Formula C8H6BrFO2
Molecular Weight (g/mol) 233.036
MDL Number MFCD09032952
InChI Key PNBIYFPZODYMOO-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorophenylacetic acid, 2-4-bromo-2-fluorophenyl acetic acid, 4-bromo-2-fluorophenyl acetic acid, 4-bromo-2-fluoro-phenyl-acetic acid, 2-4-bromo-2-fluoro-phenyl acetic acid, 4-bromo-2-fluorophenylaceticacid, pubchem13742, acmc-1bskq, ksc493s2b, 4-bromo-2-fluoro-phenylacetic acid
PubChem CID 14053792
IUPAC Name 2-(4-bromo-2-fluorophenyl)acetic acid
SMILES C1=CC(=C(C=C1Br)F)CC(=O)O

Specifications

Quantity 5 g
Formula Weight 233.03
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Bromo-2-fluorophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only