missing translation for 'onlineSavingsMsg'
Learn More

4-bromo-2-fluorobenzylamine, 98%

Catalog No. p-3212017
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC384440050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC384440050 Supplier Thermo Scientific Chemicals Supplier No. 384440050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112734-22-2 | C7H7BrFN | 204.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112734-22-2
Molecular Formula C7H7BrFN
Molecular Weight (g/mol) 204.04
MDL Number MFCD00153076
InChI Key RLTFBWCBGIZCDQ-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzylamine, 4-bromo-2-fluorophenyl methanamine, 2-fluoro-4-bromobenzylamine, 2-fluoro-4-bromobenzyl amine, 4-bromo-2-fluorobenzyl amine, benzenemethanamine, 4-bromo-2-fluoro, 1-4-bromo-2-fluorophenyl methanamine, 4-bromo-2-fluoro-phenyl methanamine, 4-bromo-2-fluorophenyl methylamine, pubchem4709
PubChem CID 3770848
IUPAC Name (4-bromo-2-fluorophenyl)methanamine
SMILES C1=CC(=C(C=C1Br)F)CN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5630 to 1.5650 (20°C, 589nm)
Linear Formula NH2CH2C6H3(F)Br
Quantity 5 g
Formula Weight 204.04
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-fluorobenzylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously

RUO – Research Use Only