accessibility menu, dialog, popup

UserName

CAS RN 112734-22-2

IUPAC Name: 1-(4-bromo-2-fluorophenyl)methanamine
Synonyms: 1-(4-bromo-2-fluorophenyl)methanamine
Molecular Weight (g/mol): 204.04
Molecular Formula: C7H7BrFN
InChi Key: RLTFBWCBGIZCDQ-UHFFFAOYSA-N
SMILES: NCC1=C(F)C=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results