missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-fluorobenzyl Chloride 98.0+%, TCI America™
$248.34 - $248.34
Chemical Identifiers
| CAS | 85510-82-3 |
|---|---|
| Molecular Formula | C7H5BrClF |
| Molecular Weight (g/mol) | 223.47 |
| MDL Number | MFCD02094036 |
| InChI Key | UDKQGFMDBMYVHI-UHFFFAOYSA-N |
| Synonym | 4-bromo-2-fluorobenzyl chloride, 4-bromo-1-chloromethyl-2-fluorobenzene, 4-bromo-2-fluorobenzylchloride, 4-bromo-1-chloromethyl-2-fluoro-benzene, acmc-209q6a, 2-fluoro-4-bromo-chloromethylbenzene |
| PubChem CID | 2773361 |
| IUPAC Name | 4-bromo-1-(chloromethyl)-2-fluorobenzene |
| SMILES | FC1=CC(Br)=CC=C1CCl |
Chemical Identifiers
| 85510-82-3 | |
| 223.47 | |
| UDKQGFMDBMYVHI-UHFFFAOYSA-N | |
| 2773361 | |
| FC1=CC(Br)=CC=C1CCl |
| C7H5BrClF | |
| MFCD02094036 | |
| 4-bromo-2-fluorobenzyl chloride, 4-bromo-1-chloromethyl-2-fluorobenzene, 4-bromo-2-fluorobenzylchloride, 4-bromo-1-chloromethyl-2-fluoro-benzene, acmc-209q6a, 2-fluoro-4-bromo-chloromethylbenzene | |
| 4-bromo-1-(chloromethyl)-2-fluorobenzene |
Specifications
| 85510-82-3 | |
| C7H5BrClF | |
| 5 g | |
| 4-bromo-2-fluorobenzyl chloride, 4-bromo-1-chloromethyl-2-fluorobenzene, 4-bromo-2-fluorobenzylchloride, 4-bromo-1-chloromethyl-2-fluoro-benzene, acmc-209q6a, 2-fluoro-4-bromo-chloromethylbenzene | |
| FC1=CC(Br)=CC=C1CCl | |
| 223.47 | |
| 223.47 | |
| Liquid |
| Yellow | |
| MFCD02094036 | |
| 3265 | |
| UDKQGFMDBMYVHI-UHFFFAOYSA-N | |
| 4-bromo-1-(chloromethyl)-2-fluorobenzene | |
| 2773361 | |
| ≥98.0% (GC) | |
| 4-Bromo-2-fluorobenzyl Chloride |
Safety and Handling
TSCA : No