accessibility menu, dialog, popup

UserName

FBrCl

CAS RN 85510-82-3

IUPAC Name: 4-bromo-1-(chloromethyl)-2-fluorobenzene
Synonyms: 4-bromo-1-(chloromethyl)-2-fluorobenzene
Molecular Weight (g/mol): 223.47
Molecular Formula: C7H5BrClF
InChi Key: UDKQGFMDBMYVHI-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC=C1CCl
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results