Learn More
4-bromo-2-fluorobenzyl Chloride, 98%, Thermo Scientific™
$59.82 - $59.82
Chemical Identifiers
| CAS | 85510-82-3 |
|---|---|
| Molecular Formula | C7H5BrClF |
| Molecular Weight (g/mol) | 223.47 |
| MDL Number | MFCD02094036 |
| InChI Key | UDKQGFMDBMYVHI-UHFFFAOYSA-N |
| Synonym | 4-bromo-2-fluorobenzyl chloride, 4-bromo-1-chloromethyl-2-fluorobenzene, 4-bromo-2-fluorobenzylchloride, 4-bromo-1-chloromethyl-2-fluoro-benzene, acmc-209q6a, 2-fluoro-4-bromo-chloromethylbenzene |
| PubChem CID | 2773361 |
| IUPAC Name | 4-bromo-1-(chloromethyl)-2-fluorobenzene |
| SMILES | FC1=CC(Br)=CC=C1CCl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC384460050
|
Thermo Scientific Chemicals
384460050 |
5 g | Glass bottle |
Each for $59.82
|
|
||||
Chemical Identifiers
| 85510-82-3 | |
| 223.47 | |
| UDKQGFMDBMYVHI-UHFFFAOYSA-N | |
| 2773361 | |
| FC1=CC(Br)=CC=C1CCl |
| C7H5BrClF | |
| MFCD02094036 | |
| 4-bromo-2-fluorobenzyl chloride, 4-bromo-1-chloromethyl-2-fluorobenzene, 4-bromo-2-fluorobenzylchloride, 4-bromo-1-chloromethyl-2-fluoro-benzene, acmc-209q6a, 2-fluoro-4-bromo-chloromethylbenzene | |
| 4-bromo-1-(chloromethyl)-2-fluorobenzene |
Specifications
| 85510-82-3 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| 1.5610 to 1.5630 (20°C, 589nm) | |
| MFCD02094036 | |
| 4-bromo-2-fluorobenzyl chloride, 4-bromo-1-chloromethyl-2-fluorobenzene, 4-bromo-2-fluorobenzylchloride, 4-bromo-1-chloromethyl-2-fluoro-benzene, acmc-209q6a, 2-fluoro-4-bromo-chloromethylbenzene | |
| FC1=CC(Br)=CC=C1CCl | |
| 223.47 | |
| 223.47 | |
| Liquid |
| 97.5 | |
| Colorless | |
| 97.5% min. (GC) | |
| C7H5BrClF | |
| ClCH2C6H3(F)Br | |
| 5 g | |
| UDKQGFMDBMYVHI-UHFFFAOYSA-N | |
| 4-bromo-1-(chloromethyl)-2-fluorobenzene | |
| 2773361 | |
| 98% | |
| 4-Bromo-2-fluorobenzyl chloride |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only