missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzoyl chloride, 98%

Catalog No. AAL2014203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2014203 1 g
AAL2014206 5 g
AAL2014214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL2014203 Supplier Thermo Scientific Chemicals Supplier No. L2014203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 151982-51-3 | C7H3BrClFO | 237.452 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 151982-51-3
Molecular Formula C7H3BrClFO
Molecular Weight (g/mol) 237.452
MDL Number MFCD03411584
InChI Key PCFIABOQFAFDAU-UHFFFAOYSA-N
PubChem CID 2734026
IUPAC Name 4-bromo-2-fluorobenzoyl chloride
SMILES C1=CC(=C(C=C1Br)F)C(=O)Cl

Specifications

Melting Point 24°C to 25°C
Boiling Point 86°C to 88°C (15 mmHg)
Refractive Index 1.5877
Quantity 1 g
UN Number UN3261
Beilstein 6323925
Sensitivity Moisture sensitive
Synonym 2-fluoro-4-bromobenzoyl chloride, benzoyl chloride, 4-bromo-2-fluoro, benzoyl chloride,4-bromo-2-fluoro, acmc-20amnm, 4-bromo-2-fluorobenzoylchloride, 4-bromo-2-fluoro-benzoyl chloride, 4-bromo-2-fluorobenzoic acid chloride, 4-bromanyl-2-fluoranyl-benzoyl chloride
Formula Weight 237.45
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-fluorobenzoyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only