accessibility menu, dialog, popup

UserName

FBrClO

CAS RN 151982-51-3

IUPAC Name: 4-bromo-2-fluorobenzoyl chloride
Synonyms: 4-bromo-2-fluorobenzoyl chloride
Molecular Weight (g/mol): 237.45
Molecular Formula: C7H3BrClFO
InChi Key: PCFIABOQFAFDAU-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC=C1C(Cl)=O
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)