Learn More
4-Bromo-2-fluorobenzoic acid, 98%
CAS: 112704-79-7 | C7H3BrFO2 | 218.00 g/mol
Supplier: Thermo Scientific Chemicals B2011814
Description
4-Bromo-2-fluorobenzoic acid may be used in the synthesis of biaryl intermediates by palladium-mediated coupling with various aryl boronic acids
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromo-2-fluorobenzoic acid may be used in the synthesis of biaryl intermediates by palladium-mediated coupling with various aryl boronic acids
Solubility
Solubility in methanol, very faint turbidity.
Notes
Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 4-Bromo-2-fluorobenzoic acid | |
| 210°C to 213°C | |
| MFCD00143263 | |
| 6323931 | |
| ZQQSRVPOAHYHEL-UHFFFAOYSA-M | |
| 4-bromo-2-fluorobenzoic acid | |
| 302681 | |
| 98% |
| 112704-79-7 | |
| C7H3BrFO2 | |
| 25 g | |
| 2-fluoro-4-bromobenzoic acid, 4-bromo-2-fluoro-benzoic acid, 4-bromo-2-fluorobenzoicacid, benzoic acid, 4-bromo-2-fluoro, pubchem1313, enzalutamide impurity a, acmc-1btmf, 2-fluro-4-bromobenzoic acid, ksc174m4p, rarechem al bo 0754 | |
| [O-]C(=O)C1=CC=C(Br)C=C1F | |
| 218.00 | |
| 219.01 |
Chemical Identifiers
| 112704-79-7 | |
| 218.00 | |
| ZQQSRVPOAHYHEL-UHFFFAOYSA-M | |
| 302681 | |
| [O-]C(=O)C1=CC=C(Br)C=C1F |
| C7H3BrFO2 | |
| MFCD00143263 | |
| 2-fluoro-4-bromobenzoic acid, 4-bromo-2-fluoro-benzoic acid, 4-bromo-2-fluorobenzoicacid, benzoic acid, 4-bromo-2-fluoro, pubchem1313, enzalutamide impurity a, acmc-1btmf, 2-fluro-4-bromobenzoic acid, ksc174m4p, rarechem al bo 0754 | |
| 4-bromo-2-fluorobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : Yes
RUO – Research Use Only