missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzoic acid, 98%

Catalog No. p-7024873
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2011806 5 g
AAB2011814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2011806 Supplier Thermo Scientific Chemicals Supplier No. B2011806

CAS: 112704-79-7 | C7H3BrFO2 | 218.00 g/mol

4-Bromo-2-fluorobenzoic acid may be used in the synthesis of biaryl intermediates by palladium-mediated coupling with various aryl boronic acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-fluorobenzoic acid may be used in the synthesis of biaryl intermediates by palladium-mediated coupling with various aryl boronic acids

Solubility
Solubility in methanol, very faint turbidity.

Notes
Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 112704-79-7
Molecular Formula C7H3BrFO2
Molecular Weight (g/mol) 218.00
MDL Number MFCD00143263
InChI Key ZQQSRVPOAHYHEL-UHFFFAOYSA-M
Synonym 2-fluoro-4-bromobenzoic acid, 4-bromo-2-fluoro-benzoic acid, 4-bromo-2-fluorobenzoicacid, benzoic acid, 4-bromo-2-fluoro, pubchem1313, enzalutamide impurity a, acmc-1btmf, 2-fluro-4-bromobenzoic acid, ksc174m4p, rarechem al bo 0754
PubChem CID 302681
IUPAC Name 4-bromo-2-fluorobenzoic acid
SMILES [O-]C(=O)C1=CC=C(Br)C=C1F

Specifications

Melting Point 210°C to 213°C
Quantity 5 g
Beilstein 6323931
Formula Weight 219.01
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-fluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA Yes

RUO – Research Use Only