missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-fluoro-1-nitrobenzene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 86°C |
| Quantity | 5 g |
| Formula Weight | 220.00 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Bromo-2-fluoro-1-nitrobenzene |
Safety and Handling
| TSCA | No |