accessibility menu, dialog, popup

UserName

BrFNOO

CAS RN 321-23-3

IUPAC Name: 4-bromo-2-fluoro-1-nitrobenzene
Synonyms: 4-bromo-2-fluoro-1-nitrobenzene
Molecular Weight (g/mol): 220
Molecular Formula: C6H3BrFNO2
InChi Key: VQCWSOYHHXXWSP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(F)C=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results