missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluoro-1-iodobenzene, 98%

Catalog No. aaa1588422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1588422 100 g
AAA1588414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1588422 Supplier Thermo Scientific Chemicals Supplier No. A1588422
Only null left

CAS: 105931-73-5 | C6H3BrFI | 300.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105931-73-5
Molecular Formula C6H3BrFI
Molecular Weight (g/mol) 300.90
MDL Number MFCD00010608
InChI Key XRMZKCQCINEBEI-UHFFFAOYSA-N
Synonym 1-bromo-3-fluoro-4-iodobenzene, 3-fluoro-4-iodobromobenzene, 2-fluoro-4-bromoiodobenzene, 4-bromo-2-fluoroiodobenzene, 4-bromo-2-fluoro-1-iodo-benzene, benzene, 4-bromo-2-fluoro-1-iodo, pubchem3232, acmc-2098hz, 2-fluoro-4-bromo iodobenzene, 3-fluoro-4-iodo-bromobenzene
PubChem CID 2724516
IUPAC Name 4-bromo-2-fluoro-1-iodobenzene
SMILES FC1=C(I)C=CC(Br)=C1

Specifications

Melting Point 46°C to 50°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 4740250
Sensitivity Light sensitive
Formula Weight 300.9
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-fluoro-1-iodobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only