missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-ethylaniline, 97%

Catalog No. AAL1272506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1272506 5 g
AAL1272514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1272506 Supplier Thermo Scientific Chemicals Supplier No. L1272506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 45762-41-2 | C8H10BrN | 200.079 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 45762-41-2
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD00041434
InChI Key LGOZNQPHTIGMQJ-UHFFFAOYSA-N
Synonym benzenamine, 4-bromo-2-ethyl, 2-ethyl-4-bromoaniline, acmc-20amnq, pubchem23174, 4-bromo-2-ethyl-aniline, 4-bromo-2-ethylbenzenamine, 4-bromo-2-ethylphenylamine, 4-bromanyl-2-ethyl-aniline, 4-bromo-2-ethylaniline
PubChem CID 2801283
IUPAC Name 4-bromo-2-ethylaniline
SMILES CCC1=C(C=CC(=C1)Br)N

Specifications

Density 1.412
Boiling Point 110°C to 112°C (2 mmHg)
Refractive Index 1.602
Quantity 5 g
UN Number UN2810
Beilstein 2802710
Formula Weight 200.08
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-ethylaniline
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only