missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-chlorobenzonitrile 97.0+%, TCI America™
Supplier: TCI America B42411G
Specifications
| 4-Bromo-2-chlorobenzonitrile | |
| 71°C | |
| 143°C | |
| MFCD00040883 | |
| 3439 | |
| AYQBMZNSJPVADT-UHFFFAOYSA-N | |
| 4-bromo-2-chlorobenzonitrile | |
| 7010420 | |
| ≥97.0% (GC) |
| 154607-01-9 | |
| White-Yellow | |
| C7H3BrClN | |
| 1 g | |
| 2-chloro-4-bromobenzonitrile, 4-bromo-2-chloro-benzonitrile, benzonitrile, 4-bromo-2-chloro, 4-bromo-2-chlorobenzenecarbonitrile, pubchem12801, intermediates-zcf02177, acmc-209db5, ksc497g8d | |
| ClC1=C(C=CC(Br)=C1)C#N | |
| 216.46 | |
| 216.46 | |
| Crystalline Powder |
Chemical Identifiers
| 154607-01-9 | |
| 216.46 | |
| AYQBMZNSJPVADT-UHFFFAOYSA-N | |
| 7010420 | |
| ClC1=C(C=CC(Br)=C1)C#N |
| C7H3BrClN | |
| MFCD00040883 | |
| 2-chloro-4-bromobenzonitrile, 4-bromo-2-chloro-benzonitrile, benzonitrile, 4-bromo-2-chloro, 4-bromo-2-chlorobenzenecarbonitrile, pubchem12801, intermediates-zcf02177, acmc-209db5, ksc497g8d | |
| 4-bromo-2-chlorobenzonitrile |
Safety and Handling
TSCA : No