missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chlorobenzonitrile 97.0+%, TCI America™

Catalog No. p-7123425
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42411G 1 g
B42415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42411G Supplier TCI America Supplier No. B42411G
Only null left

Chemical Identifiers

CAS 154607-01-9
Molecular Formula C7H3BrClN
Molecular Weight (g/mol) 216.46
MDL Number MFCD00040883
InChI Key AYQBMZNSJPVADT-UHFFFAOYSA-N
Synonym 2-chloro-4-bromobenzonitrile, 4-bromo-2-chloro-benzonitrile, benzonitrile, 4-bromo-2-chloro, 4-bromo-2-chlorobenzenecarbonitrile, pubchem12801, intermediates-zcf02177, acmc-209db5, ksc497g8d
PubChem CID 7010420
IUPAC Name 4-bromo-2-chlorobenzonitrile
SMILES ClC1=C(C=CC(Br)=C1)C#N

Specifications

Melting Point 71°C
Color White-Yellow
Boiling Point 143°C
Quantity 1 g
UN Number 3439
Formula Weight 216.46
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-chlorobenzonitrile