missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloroaniline 98.0+%, TCI America™

$59.69 - $200.21

Chemical Identifiers

CAS 38762-41-3
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD00007660
InChI Key INMZDDDQLHKGPF-UHFFFAOYSA-N
Synonym 2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline
PubChem CID 610169
IUPAC Name 4-bromo-2-chloroaniline
SMILES C1=CC(=C(C=C1Br)Cl)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B19935G
View Documents
TCI America
B19935G
5 g
Each for $59.69
Only null left
 
B199325G
View Documents
TCI America
B199325G
25 g
Each for $200.21
Only null left
 
Description

Description

Specifications

Chemical Identifiers

38762-41-3
206.467
INMZDDDQLHKGPF-UHFFFAOYSA-N
610169
C1=CC(=C(C=C1Br)Cl)N
C6H5BrClN
MFCD00007660
2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline
4-bromo-2-chloroaniline

Specifications

38762-41-3
C6H5BrClN
5 g
INMZDDDQLHKGPF-UHFFFAOYSA-N
4-bromo-2-chloroaniline
610169
≥98.0% (GC)
4-Bromo-2-chloroaniline
70°C
MFCD00007660
2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline
C1=CC(=C(C=C1Br)Cl)N
206.467
206.47
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents