missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-chloroaniline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 70°C |
| Quantity | 25 g |
| Formula Weight | 206.47 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Bromo-2-chloroaniline |
Safety and Handling
| TSCA | No |