missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloro-1-iodobenzene, 97%

Catalog No. p-7026269
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2218503 1 g
AAB2218506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2218503 Supplier Thermo Scientific Chemicals Supplier No. B2218503
Only null left

CAS: 31928-47-9 | C6H3BrClI | 317.348 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31928-47-9
Molecular Formula C6H3BrClI
Molecular Weight (g/mol) 317.348
MDL Number MFCD00079709
InChI Key OHHKQBZOURGNLR-UHFFFAOYSA-N
Synonym 4-bromo-2-chloroiodobenzene, 1-bromo-3-chloro-4-iodobenzene, benzene, 4-bromo-2-chloro-1-iodo, buttpark 34\07-78, 4-bromo-2-chloro-1-iodo-benzene, pubchem3592, acmc-1cp6y, 3-chloro-4-iodobromobenzene, ksc493m0p, 4-bromo-2-chloro-iodobenzene
PubChem CID 2735547
IUPAC Name 4-bromo-2-chloro-1-iodobenzene
SMILES C1=CC(=C(C=C1Br)Cl)I

Specifications

Melting Point 33°C to 35°C
Boiling Point 103°C to 105°C (1 mmHg)
Refractive Index 1.6722
Quantity 1 g
Beilstein 2435075
Sensitivity Light sensitive
Formula Weight 317.36
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-chloro-1-iodobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only