missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloro-1-fluorobenzene 98.0+%, TCI America™

Catalog No. B18605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18605G 5 g
B186025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18605G Supplier TCI America Supplier No. B18605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60811-21-4
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.44
MDL Number MFCD00051794
InChI Key CJTIWGBQCVYTQE-UHFFFAOYSA-N
Synonym 1-bromo-3-chloro-4-fluorobenzene, 3-chloro-4-fluorobromobenzene, 4-bromo-2-chlorofluorobenzene, 3-chloro-4-fluorobenzene bromide, 4-bromo-2-chloro-fluorobenzene, 4-bromo-2-chloro-1-fluoro-benzene, benzene, 4-bromo-2-chloro-1-fluoro, pubchem2169, acmc-209mma, ksc493o4n
PubChem CID 3543065
IUPAC Name 4-bromo-2-chloro-1-fluorobenzene
SMILES FC1=C(Cl)C=C(Br)C=C1

Specifications

Color Yellow
Boiling Point 95°C
Quantity 5 g
UN Number 3082
Formula Weight 209.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-chloro-1-fluorobenzene