accessibility menu, dialog, popup

UserName

BrClF

CAS RN 60811-21-4

IUPAC Name: 4-bromo-2-chloro-1-fluorobenzene
Synonyms: 4-bromo-2-chloro-1-fluorobenzene
Molecular Weight (g/mol): 209.44
Molecular Formula: C6H3BrClF
InChi Key: CJTIWGBQCVYTQE-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=C(Br)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)