missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloro-1-fluorobenzene, 98+%

Catalog No. p-7024679
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1978214 25 g
AAA1978222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1978214 Supplier Thermo Scientific Chemicals Supplier No. A1978214

CAS: 60811-21-4 | C6H3BrClF | 209.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60811-21-4
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.44
MDL Number MFCD00051794
InChI Key CJTIWGBQCVYTQE-UHFFFAOYSA-N
Synonym 1-bromo-3-chloro-4-fluorobenzene, 3-chloro-4-fluorobromobenzene, 4-bromo-2-chlorofluorobenzene, 3-chloro-4-fluorobenzene bromide, 4-bromo-2-chloro-fluorobenzene, 4-bromo-2-chloro-1-fluoro-benzene, benzene, 4-bromo-2-chloro-1-fluoro, pubchem2169, acmc-209mma, ksc493o4n
PubChem CID 3543065
IUPAC Name 4-bromo-2-chloro-1-fluorobenzene
SMILES FC1=C(Cl)C=C(Br)C=C1

Specifications

Density 1.727
Boiling Point 188°C to 190°C
Flash Point 88°C (190°F)
Refractive Index 1.554
Quantity 25 g
UN Number UN3082
Beilstein 2206652
Formula Weight 209.45
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-2-chloro-1-fluorobenzene
Hazard Category H227-H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H227
Harmful if swallowed.
Causes skin irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 405-580-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only