missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2,6-pyridinedicarboxylic Acid Monohydrate 98.0+%, TCI America™
$309.43 - $1053.19
Chemical Identifiers
| CAS | 162102-81-0 |
|---|---|
| Molecular Formula | C7H4BrNO4 |
| Molecular Weight (g/mol) | 246.016 |
| MDL Number | MFCD09033573 |
| InChI Key | JVCWTTKJXQPITQ-UHFFFAOYSA-N |
| PubChem CID | 15832860 |
| IUPAC Name | 4-bromopyridine-2,6-dicarboxylic acid |
| SMILES | C1=C(C=C(N=C1C(=O)O)C(=O)O)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4970200MG
|
TCI America
B4970200MG |
200 mg |
Each for $309.43
|
|
|||||
|
B49701G
|
TCI America
B49701G |
1 g |
Each for $1,053.19
|
|
|||||
Chemical Identifiers
| 162102-81-0 | |
| 246.016 | |
| JVCWTTKJXQPITQ-UHFFFAOYSA-N | |
| 4-bromopyridine-2,6-dicarboxylic acid |
| C7H4BrNO4 | |
| MFCD09033573 | |
| 15832860 | |
| C1=C(C=C(N=C1C(=O)O)C(=O)O)Br |
Specifications
| 162102-81-0 | |
| White | |
| MFCD09033573 | |
| JVCWTTKJXQPITQ-UHFFFAOYSA-N | |
| 4-bromopyridine-2,6-dicarboxylic acid | |
| 15832860 | |
| ≥98.0% (GC,T) | |
| 4-Bromo-2,6-pyridinedicarboxylic Acid Monohydrate |
| 206°C | |
| C7H4BrNO4 | |
| 200 mg | |
| C1=C(C=C(N=C1C(=O)O)C(=O)O)Br | |
| 246.016 | |
| 246.02 | |
| Crystalline Powder |
Safety and Handling
TSCA : No