missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2,6-pyridinedicarboxylic Acid Monohydrate 98.0+%, TCI America™
Supplier: TCI America B49701G
Specifications
| 4-Bromo-2,6-pyridinedicarboxylic Acid Monohydrate | |
| 206°C | |
| C7H4BrNO4 | |
| 1 g | |
| C1=C(C=C(N=C1C(=O)O)C(=O)O)Br | |
| 246.016 | |
| 246.02 | |
| Crystalline Powder |
| 162102-81-0 | |
| White | |
| MFCD09033573 | |
| JVCWTTKJXQPITQ-UHFFFAOYSA-N | |
| 4-bromopyridine-2,6-dicarboxylic acid | |
| 15832860 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 162102-81-0 | |
| 246.016 | |
| JVCWTTKJXQPITQ-UHFFFAOYSA-N | |
| 4-bromopyridine-2,6-dicarboxylic acid |
| C7H4BrNO4 | |
| MFCD09033573 | |
| 15832860 | |
| C1=C(C=C(N=C1C(=O)O)C(=O)O)Br |
Safety and Handling
TSCA : No