missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-dimethylphenol 98.0+%, TCI America™

Catalog No. B206025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B206025G 25 g
B2060250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B206025G Supplier TCI America Supplier No. B206025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2374-05-2
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00002314
InChI Key ZLVFYUORUHNMBO-UHFFFAOYSA-N
Synonym 4-bromo-2,6-xylenol, phenol, 4-bromo-2,6-dimethyl, 2,6-dimethyl-4-bromophenol, 2,6-xylenol, 4-bromo, pubchem9579, 2, 4-bromo, acmc-2097aq, 4-bromo-2,6 dimethylphenol, 5-bromo-2-hydroxy-m-xylene, ksc495g0d
PubChem CID 16919
IUPAC Name 4-bromo-2,6-dimethylphenol
SMILES CC1=CC(=CC(=C1O)C)Br

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 25 g
Formula Weight 201.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,6-dimethylphenol
RTECSNumber ZE6780000
TSCA Yes