accessibility menu, dialog, popup

UserName

CAS RN 2374-05-2

IUPAC Name: 4-bromo-2,6-dimethylphenol
Synonyms: phenol, 4-bromo-2,6-dimethyl-
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: ZLVFYUORUHNMBO-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC(C)=C1O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)