missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-dimethylphenol, 99%

Catalog No. AAB2166914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2166914 25 g
AAB2166922 100 g
AAB2166936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2166914 Supplier Thermo Scientific Chemicals Supplier No. B2166914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2374-05-2 | C8H9BrO | 201.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It is used as a preservatives for coatings, slurries and to control microbial fouling in paper mills , oil field and leather process and water treatment process against microbial attack.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 2374-05-2
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00002314
InChI Key ZLVFYUORUHNMBO-UHFFFAOYSA-N
Synonym 4-bromo-2,6-xylenol, phenol, 4-bromo-2,6-dimethyl, 2,6-dimethyl-4-bromophenol, 2,6-xylenol, 4-bromo, pubchem9579, 2, 4-bromo, acmc-2097aq, 4-bromo-2,6 dimethylphenol, 5-bromo-2-hydroxy-m-xylene, ksc495g0d
PubChem CID 16919
IUPAC Name 4-bromo-2,6-dimethylphenol
SMILES CC1=CC(=CC(=C1O)C)Br

Specifications

Melting Point 74°C to 78°C
Odor Phenol-like
Quantity 25 g
Beilstein 1862399
Solubility Information Slightly soluble in water.
Formula Weight 201.07
Percent Purity 99%
Chemical Name or Material 4-Bromo-2,6-dimethylphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 219-153-1
RTECSNumber ZE6780000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only