missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2,6-dimethylanisole 96.0+%, TCI America™
$71.23 - $219.48
Chemical Identifiers
| CAS | 14804-38-7 |
|---|---|
| Molecular Formula | C9H11BrO |
| Molecular Weight (g/mol) | 215.09 |
| MDL Number | MFCD00143259 |
| InChI Key | MMARFGDTMJBIBK-UHFFFAOYSA-N |
| Synonym | 4-bromo-2,6-dimethylanisole, 5-bromo-2-methoxy-m-xylene, 4-brom-2,6-dimethylanisole, 1-bromo-3,5-dimethyl-4-methoxybenzene, 2,6-dimethyl-4-bromoanisole, benzene, 5-bromo-2-methoxy-1,3-dimethyl, timtec-bb sbb005776, attercop-chm at108629, 4-bromo-2,6-dimethyl-methoxybenzene, pubchem2654 |
| PubChem CID | 278654 |
| IUPAC Name | 5-bromo-2-methoxy-1,3-dimethylbenzene |
| SMILES | COC1=C(C)C=C(Br)C=C1C |
Chemical Identifiers
| 14804-38-7 | |
| 215.09 | |
| MMARFGDTMJBIBK-UHFFFAOYSA-N | |
| 278654 | |
| COC1=C(C)C=C(Br)C=C1C |
| C9H11BrO | |
| MFCD00143259 | |
| 4-bromo-2,6-dimethylanisole, 5-bromo-2-methoxy-m-xylene, 4-brom-2,6-dimethylanisole, 1-bromo-3,5-dimethyl-4-methoxybenzene, 2,6-dimethyl-4-bromoanisole, benzene, 5-bromo-2-methoxy-1,3-dimethyl, timtec-bb sbb005776, attercop-chm at108629, 4-bromo-2,6-dimethyl-methoxybenzene, pubchem2654 | |
| 5-bromo-2-methoxy-1,3-dimethylbenzene |
Specifications
| 14804-38-7 | |
| 120°C | |
| Clear Liquid at 20°C | |
| MFCD00143259 | |
| MMARFGDTMJBIBK-UHFFFAOYSA-N | |
| 5-bromo-2-methoxy-1,3-dimethylbenzene | |
| 278654 | |
| ≥96.0% (GC) |
| Yellow | |
| 5 g | |
| C9H11BrO | |
| 4-bromo-2,6-dimethylanisole, 5-bromo-2-methoxy-m-xylene, 4-brom-2,6-dimethylanisole, 1-bromo-3,5-dimethyl-4-methoxybenzene, 2,6-dimethyl-4-bromoanisole, benzene, 5-bromo-2-methoxy-1,3-dimethyl, timtec-bb sbb005776, attercop-chm at108629, 4-bromo-2,6-dimethyl-methoxybenzene, pubchem2654 | |
| COC1=C(C)C=C(Br)C=C1C | |
| 215.09 | |
| 215.09 | |
| 4-Bromo-2,6-dimethylanisole |
Safety and Handling
TSCA : No