accessibility menu, dialog, popup

UserName

H3COCH3BrH3C

CAS RN 14804-38-7

IUPAC Name: 5-bromo-2-methoxy-1,3-dimethylbenzene
Synonyms: 5-bromo-2-methoxy-1,3-dimethylbenzene
Molecular Weight (g/mol): 215.09
Molecular Formula: C9H11BrO
InChi Key: MMARFGDTMJBIBK-UHFFFAOYSA-N
SMILES: COC1=C(C)C=C(Br)C=C1C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)