missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2,6-dimethylanisole 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| Physical Form | Clear Liquid at 20°C |
| Boiling Point | 120°C |
| Formula Weight | 215.09 |
| Percent Purity | ≥96.0% (GC) |
| Grade | Not Available |
| Chemical Name or Material | 4-Bromo-2,6-dimethylanisole |
Safety and Handling
| TSCA | No |