missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-difluorobenzyl Alcohol 98.0+%, TCI America™

Catalog No. p-7123944
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50901G 1 g
B50905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50901G Supplier TCI America Supplier No. B50901G

Chemical Identifiers

CAS 162744-59-4
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.017
MDL Number MFCD03094461
InChI Key LSRHFWSNUFIKER-UHFFFAOYSA-N
Synonym 4-bromo-2,6-difluorobenzyl alcohol, 4-bromo-2,6-difluorophenyl methanol, 4-bromo-2,6-difluorobenzylalcohol, benzenemethanol, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluoro-benzenemethanol, 2,6-difluoro-4-bromobenzyl alcohol, 4-bromo-2,6-difluoro-phenyl methanol, 2,6-difluoro-4-bromo benzyl alcohol, pubchem4882, acmc-209dox
PubChem CID 2773303
IUPAC Name (4-bromo-2,6-difluorophenyl)methanol
SMILES C1=C(C=C(C(=C1F)CO)F)Br

Specifications

Melting Point 81°C
Color White-Yellow
Quantity 1 g
Formula Weight 223.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,6-difluorobenzyl Alcohol