accessibility menu, dialog, popup

UserName

FOHFBr

CAS RN 162744-59-4

IUPAC Name: (4-bromo-2,6-difluorophenyl)methanol
Synonyms: (4-bromo-2,6-difluorophenyl)methanol
Molecular Weight (g/mol): 223.02
Molecular Formula: C7H5BrF2O
InChi Key: LSRHFWSNUFIKER-UHFFFAOYSA-N
SMILES: OCC1=C(F)C=C(Br)C=C1F
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)