missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-difluorobenzyl alcohol, 97%

Catalog No. p-7053878
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6162303 1 g
AAH6162306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6162303 Supplier Thermo Scientific Chemicals Supplier No. H6162303
Only null left

CAS: 162744-59-4 | C7H5BrF2O | 223.017 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 162744-59-4
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.017
MDL Number MFCD03094461
InChI Key LSRHFWSNUFIKER-UHFFFAOYSA-N
Synonym 4-bromo-2,6-difluorobenzyl alcohol, 4-bromo-2,6-difluorophenyl methanol, 4-bromo-2,6-difluorobenzylalcohol, benzenemethanol, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluoro-benzenemethanol, 2,6-difluoro-4-bromobenzyl alcohol, 4-bromo-2,6-difluoro-phenyl methanol, 2,6-difluoro-4-bromo benzyl alcohol, pubchem4882, acmc-209dox
PubChem CID 2773303
IUPAC Name (4-bromo-2,6-difluorophenyl)methanol
SMILES C1=C(C=C(C(=C1F)CO)F)Br

Specifications

Melting Point 76°C to 81°C
Quantity 1 g
Formula Weight 223.01
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Bromo-2,6-difluorobenzyl alcohol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only