missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-difluorobenzotrifluoride, 98%

CAS: 156243-64-0 | C7H2BrF5 | 260.989 g/mol

$115.47 - $404.26

Chemical Identifiers

CAS 156243-64-0
Molecular Formula C7H2BrF5
Molecular Weight (g/mol) 260.989
MDL Number MFCD01631616
InChI Key QPJKIRNIIXIPIE-UHFFFAOYSA-N
PubChem CID 23450919
IUPAC Name 5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene
SMILES C1=C(C=C(C(=C1F)C(F)(F)F)F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6438603
View Documents
Thermo Scientific Chemicals
H6438603
1 g
Each for $115.47
Only null left
 
AAH6438606
View Documents
Thermo Scientific Chemicals
H6438606
5 g
Each for $404.26
Only null left
 
Description

Description

It is employed in the lewis acid-promoted synthesis of unsymmetrical and highly functionalized carbazoles and dibenzofurans from biaryl triazenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the lewis acid-promoted synthesis of unsymmetrical and highly functionalized carbazoles and dibenzofurans from biaryl triazenes.

Solubility
Slightly soluble in water.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat.
Specifications

Chemical Identifiers

156243-64-0
260.989
QPJKIRNIIXIPIE-UHFFFAOYSA-N
5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene
C7H2BrF5
MFCD01631616
23450919
C1=C(C=C(C(=C1F)C(F)(F)F)F)Br

Specifications

156243-64-0
C7H2BrF5
1 g
3,5-difluoro-4-trifluoromethyl bromobenzene, 5-bromo-1,3-difluoro-2-trifluoromethyl benzene, 4-bromo-2,6-difluorobenzotrifluoride, benzene, 5-bromo-1,3-difluoro-2-trifluoromethyl, acmc-1cioh, intermediates-zcf02186, 2,6-difluoro-4-bromobenzotrifluoride, 1-bromo-3,5-difluoro-4-trifluoromethylbenzene, 3,5-difluoro-4-trifluoromethylbromobenzene, benzene,5-bromo-1,3-difluoro-2-trifluoromethyl
QPJKIRNIIXIPIE-UHFFFAOYSA-N
5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene
23450919
98%
4-Bromo-2,6-difluorobenzotrifluoride
Colorless
MFCD01631616
13252396
Slightly soluble in water.
C1=C(C=C(C(=C1F)C(F)(F)F)F)Br
260.989
260.99
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only