accessibility menu, dialog, popup

UserName

CAS RN 156243-64-0

FFFFFBr

CAS RN 156243-64-0

IUPAC Name: 5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene
Synonyms: 5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 260.99
Molecular Formula: C7H2BrF5
InChi Key: QPJKIRNIIXIPIE-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC(F)=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results