missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-difluorobenzonitrile 98.0+%, TCI America™

Catalog No. B40215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40215G 5 g
B40211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40215G Supplier TCI America Supplier No. B40215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123843-67-4
Molecular Formula C7H2BrF2N
Molecular Weight (g/mol) 218.00
MDL Number MFCD01310981
InChI Key TZHQWUAOIWRFSW-UHFFFAOYSA-N
Synonym 4-bromo-2,6-difluoro-benzonitrile, 2,6-difluoro-4-bromobenzonitrile, benzonitrile, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluorobenzenecarbonitrile, pubchem4699, acmc-1bymk, ksc513m5d, 4-bromo-2,6-difluoro-bezonitrile, 2,6-difluoro-4-bromo-benzonitrile
PubChem CID 2756916
IUPAC Name 4-bromo-2,6-difluorobenzonitrile
SMILES FC1=CC(Br)=CC(F)=C1C#N

Specifications

Melting Point 81°C
Color White
Quantity 5 g
UN Number 3439
Formula Weight 218.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,6-difluorobenzonitrile