accessibility menu, dialog, popup

UserName

FNFBr

CAS RN 123843-67-4

IUPAC Name: 4-bromo-2,6-difluorobenzonitrile
Synonyms: 4-bromo-2,6-difluorobenzonitrile
Molecular Weight (g/mol): 218
Molecular Formula: C7H2BrF2N
InChi Key: TZHQWUAOIWRFSW-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC(F)=C1C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results