missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-dichlorophenol, 97%

Catalog No. AAH6676306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6676306 5 g
AAH6676303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6676306 Supplier Thermo Scientific Chemicals Supplier No. H6676306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3217-15-0 | C6H3BrCl2O | 241.893 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3217-15-0
Molecular Formula C6H3BrCl2O
Molecular Weight (g/mol) 241.893
MDL Number MFCD00019996
InChI Key KGURSDWHGSLAPP-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-bromophenol, 4-bromo-2.6-dichlorophenol, 4-bromo-2 6-dichlorophenol, 4-bromo-2,6-dichloro-phenol, phenol, 4-bromo-2,6-dichloro, pubchem3908, acmc-209hrm, ksc568g3h, phenol,4-bromo-2,6-dichloro, phenol, 4-bromo-2,6-dichloro-8ci 9ci
PubChem CID 18587
IUPAC Name 4-bromo-2,6-dichlorophenol
SMILES C1=C(C=C(C(=C1Cl)O)Cl)Br

Specifications

Color White
Quantity 5 g
UN Number UN2811
Beilstein 2046857
Formula Weight 241.89
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Bromo-2,6-dichlorophenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1
EINECSNumber 221-740-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only