missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2,6-di-tert-butylphenol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 83°C |
| Boiling Point | 128°C |
| Quantity | 25 g |
| Formula Weight | 285.23 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Bromo-2,6-di-tert-butylphenol |
Safety and Handling
| TSCA | No |